siRNA Details
Gene Symbol: CXorf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203407.2 | NP_981952.1 | 1 | 862 | ||
GenBank mRNA | BC046248.1 | - | 1 | 862 | 6046539 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CXorf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203407.2 | NP_981952.1 | 1 | 862 | ||
GenBank mRNA | BC046248.1 | - | 1 | 862 | 6046539 |