siRNA Details
| Gene Symbol: DCDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181807.3 | NP_861523.2 | 3 | 277 | ||
| GenBank mRNA | AY247970.1 | - | 3 | 241 | ||
| DB090201.1 | - | 3 | 277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCDC1| Gene Symbol: DCDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181807.3 | NP_861523.2 | 3 | 277 | ||
| GenBank mRNA | AY247970.1 | - | 3 | 241 | ||
| DB090201.1 | - | 3 | 277 | |||