siRNA Details
| Gene Symbol: C11orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198498.2 | NP_940900.1 | 3, 4 | 449 | ||
| XM_011542803.2 | XP_011541105.1 | 4, 5 | 592 | |||
| XM_011542804.2 | XP_011541106.1 | 3, 4 | 375 | |||
| GenBank mRNA | BC039669.1 | - | 3, 4 | 449 | 5167284 | |

C11orf53
Genomic Map