siRNA Details
Gene Symbol: OR2AT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005285.1 | NP_001005285.1 | 1 | 539 | ||
XM_017017655.1 | XP_016873144.1 | 2 | 2179 | |||
XM_017017656.1 | XP_016873145.1 | 2 | 2175 | |||
XM_017017657.1 | XP_016873146.1 | 2 | 1717 | |||
XM_017017658.1 | XP_016873147.1 | 2 | 2202 | |||
GenBank mRNA | BC136958.1 | - | 1 | 580 | 9020950 | |
BC136959.1 | - | 1 | 580 | 9020951 |
