siRNA Details
| Gene Symbol: LRRC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_075071.1 | 23 | 3302 | |||
| GenBank mRNA | AK128037.1 | - | 23 | 3302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC9| Gene Symbol: LRRC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_075071.1 | 23 | 3302 | |||
| GenBank mRNA | AK128037.1 | - | 23 | 3302 | ||