siRNA Details
| Gene Symbol: CCDC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306076.1 | NP_001293005.1 | 13 | 1949 | ||
| NM_206886.4 | NP_996769.3 | 13 | 1952 | |||
| XM_011541361.2 | XP_011539663.2 | 13 | 2161 | |||
| XM_011541372.2 | XP_011539674.2 | 13 | 2161 | |||
| XM_017001154.1 | XP_016856643.1 | 13 | 2161 | |||
| XM_017001155.1 | XP_016856644.1 | 13 | 2158 | |||
| XM_017001156.1 | XP_016856645.1 | 13 | 2131 | |||
| XM_017001157.1 | XP_016856646.1 | 13 | 2113 | |||
| XM_017001158.1 | XP_016856647.1 | 13 | 2158 | |||
| XM_017001159.1 | XP_016856648.1 | 13 | 2161 | |||
| XM_017001160.1 | XP_016856649.1 | 13 | 2161 | |||
| XM_017001161.1 | XP_016856650.1 | 13 | 2161 | |||
| XM_017001162.1 | XP_016856651.1 | 13 | 1874 | |||
| XM_017001163.1 | XP_016856652.1 | 13 | 2428 | |||
| XM_017001164.1 | XP_016856653.1 | 13 | 1972 | |||
| XM_017001165.1 | XP_016856654.1 | 13 | 2161 | |||
| XM_017001166.1 | XP_016856655.1 | 11 | 1616 | |||
| XM_017001167.1 | XP_016856656.1 | 11 | 1551 | |||
| XM_017001168.1 | XP_016856657.1 | 9 | 1316 | |||
| XM_017001169.1 | XP_016856658.1 | 13 | 2161 | |||
| GenBank mRNA | AK126045.1 | - | 7 | 1020 | ||
| BX538286.1 | - | 6 | 943 | |||

CCDC18
Genomic Map