siRNA Details
| Gene Symbol: OR2W3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001957.2 | NP_001001957.2 | 1 | 21 | ||
| GenBank mRNA | BC146848.1 | - | 1 | 52 | 9056776 | |
| Gene Symbol: TRIM58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537432.1 | - | 3 | 278 | ||

OR2W3
Genomic Map