siRNA Details
| Gene Symbol: KIAA1211L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207362.2 | NP_997245.2 | 2 | 325 | ||
| XM_005263923.3 | XP_005263980.1 | 2 | 305 | |||
| XM_005263924.3 | XP_005263981.1 | 2 | 305 | |||
| XM_005263926.3 | XP_005263983.1 | 2 | 190 | |||
| XM_011511093.1 | XP_011509395.1 | 4 | 464 | |||
| XM_011511094.1 | XP_011509396.1 | 3 | 362 | |||
| XM_011511095.2 | XP_011509397.1 | 2 | 132 | |||
| XM_017003971.1 | XP_016859460.1 | 2 | 308 | |||
| XM_017003972.1 | XP_016859461.1 | 3 | 412 | |||
| XM_017003973.1 | XP_016859462.1 | 2 | 318 | |||
| GenBank mRNA | BM922097.1 | - | 2 | 343 | ||

KIAA1211L
Genomic Map