siRNA Details
| Gene Symbol: AADACL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207365.3 | NP_997248.2 | 4 | 577 | ||
| XM_011512753.1 | XP_011511055.1 | 4 | 605 | |||
| GenBank mRNA | BC065724.1 | - | 4 | 570 | 6663150 | |
| BX647585.1 | - | 3 | 354 | |||
| CU449274.1 | - | 4 | 399 | |||

AADACL2
Genomic Map