siRNA Details
| Gene Symbol: PRSS48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183375.2 | NP_899231.2 | 5 | 885 | ||
| XM_011531921.2 | XP_011530223.1 | 4 | 1331 | |||
| XM_011531922.2 | XP_011530224.1 | 3 | 1147 | |||
| GenBank mRNA | BC121109.1 | - | 5 | 889 | 40120369 | |
| BC121110.1 | - | 3 | 460 | 40120371 | ||

PRSS48
Genomic Map