siRNA Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182762.3 | NP_877439.3 | 5 | 2217 | ||
| GenBank mRNA | AJ313524.1 | - | 2 | 1908 | ||
| AK128850.1 | - | 1 | 1378 | |||
| AK131400.1 | - | 5 | 2217 | |||
| BC137090.1 | - | 3 | 1938 | 9021087 | ||

MACC1
Genomic Map