siRNA Details
| Gene Symbol: STRA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182489.1 | NP_872295.1 | 3 | 274 | ||
| XM_011516137.2 | XP_011514439.1 | 3 | 914 | |||
| XM_011516138.2 | XP_011514440.1 | 3 | 299 | |||
| XM_011516139.2 | XP_011514441.1 | 4 | 543 | |||
| XM_017012150.1 | XP_016867639.1 | 3 | 914 | |||
| GenBank mRNA | AF513502.1 | - | 3 | 274 | ||

STRA8
Genomic Map