siRNA Details
| Gene Symbol: IGFBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007563.2 | NP_001007564.1 | 4 | 805 | ||
| GenBank mRNA | AK131521.1 | - | 4 | 358 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGFBPL1| Gene Symbol: IGFBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007563.2 | NP_001007564.1 | 4 | 805 | ||
| GenBank mRNA | AK131521.1 | - | 4 | 358 | ||