siRNA Details
| Gene Symbol: LRRTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178839.4 | NP_849161.2 | 2 | 1734 | ||
| XM_017003986.1 | XP_016859475.1 | 2 | 2266 | |||
| XM_017003987.1 | XP_016859476.1 | 2 | 2114 | |||
| GenBank mRNA | AA977181.1 | - | 1 | 480 | ||
| AK056644.1 | - | 2 | 1765 | |||
| AK290512.1 | - | 2 | 1984 | |||
| AY182024.1 | - | 1 | 1464 | |||
| AY358310.1 | - | 2 | 2183 | |||
| BC045113.1 | - | 2 | 1734 | 5200499 | ||

LRRTM1
Genomic Map