siRNA Details
| Gene Symbol: LRRTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178011.4 | NP_821079.3 | 2 | 1510 | ||
| GenBank mRNA | AK290168.1 | - | 2 | 1043 | ||
| AY182028.1 | - | 1 | 960 | |||
| AY358315.1 | - | 2 | 1467 | |||
| BC111492.1 | - | 2 | 1462 | 5196989 | ||
| BC113715.1 | - | 2 | 1020 | 8322767 | ||
| BC113717.1 | - | 2 | 1020 | 8322769 | ||
| BX640611.1 | - | 2 | 1489 | |||

LRRTM3
Genomic Map