siRNA Details
Gene Symbol: ZNF713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182633.2 | NP_872439.2 | 7 | 2129 | ||
GenBank mRNA | AI274573.1 | - | 1 | 157 | ||
AK097282.1 | - | 4 | 1142 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182633.2 | NP_872439.2 | 7 | 2129 | ||
GenBank mRNA | AI274573.1 | - | 1 | 157 | ||
AK097282.1 | - | 4 | 1142 |