siRNA Details
| Gene Symbol: ZNF429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001415.2 | NP_001001415.2 | 4 | 620 | ||
| XM_017026748.1 | XP_016882237.1 | 5 | 1119 | |||
| XM_017026749.1 | XP_016882238.1 | 5 | 3033 | |||
| XM_017026750.1 | XP_016882239.1 | 5 | 1223 | |||
| XM_017026751.1 | XP_016882240.1 | 4 | 1134 | |||
| XM_017026752.1 | XP_016882241.1 | 4 | 676 | |||
| XM_017026753.1 | XP_016882242.1 | 6 | 972 | |||
| XM_017026754.1 | XP_016882243.1 | 5 | 917 | |||
| XM_017026755.1 | XP_016882244.1 | 6 | 3056 | |||
| XM_017026756.1 | XP_016882245.1 | 5 | 924 | |||
| XM_017026757.1 | XP_016882246.1 | 3 | 870 | |||
| GenBank mRNA | AF325191.1 | - | 4 | 596 | ||
| AY269786.1 | - | 4 | 596 | |||

ZNF429
Genomic Map