siRNA Details
| Gene Symbol: TMPRSS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182973.1 | NP_892018.1 | 4 | 437 | ||
| XM_011527978.2 | XP_011526280.1 | 6 | 800 | |||
| GenBank mRNA | AB109390.1 | - | 5 | 554 | ||
| AJ488946.1 | - | 4 | 437 | |||
| AJ488947.1 | - | 4 | 437 | |||

TMPRSS9
Genomic Map