siRNA Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182974.2 | NP_892019.2 | 5 | 1135 | ||
| XM_011518633.1 | XP_011516935.1 | 8 | 1459 | |||
| XM_011518634.1 | XP_011516936.1 | 7 | 1398 | |||
| XM_011518635.1 | XP_011516937.1 | 8 | 1364 | |||
| XM_011518636.1 | XP_011516938.1 | 7 | 1303 | |||
| XM_011518637.1 | XP_011516939.1 | 7 | 1351 | |||
| XM_011518638.2 | XP_011516940.1 | 5 | 1061 | |||
| GenBank mRNA | AY336054.1 | - | 5 | 1135 | ||

GLT6D1
Genomic Map