siRNA Details
| Gene Symbol: PRSS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135086.1 | NP_001128558.1 | 3 | 378 | ||
| XM_011522477.2 | XP_011520779.1 | 5 | 996 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS41| Gene Symbol: PRSS41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135086.1 | NP_001128558.1 | 3 | 378 | ||
| XM_011522477.2 | XP_011520779.1 | 5 | 996 | |||