siRNA Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198514.3 | NP_940916.2 | 3 | 638 | ||
| XM_011539769.2 | XP_011538071.1 | 3 | 638 | |||
| GenBank mRNA | AK090631.1 | - | 3 | 609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NHLRC2| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198514.3 | NP_940916.2 | 3 | 638 | ||
| XM_011539769.2 | XP_011538071.1 | 3 | 638 | |||
| GenBank mRNA | AK090631.1 | - | 3 | 609 | ||