siRNA Details
| Gene Symbol: LOC100287896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319240.1 | NP_001306169.1 | 2 | 1466 | ||
| XM_017017055.1 | XP_016872544.1 | 2 | 954 | |||
| GenBank mRNA | BC019340.1 | - | 2 | 1411 | 3051084 | |
| BX647206.1 | - | 2 | 1055 | |||

LOC100287896
Genomic Map