siRNA Details
Gene Symbol: HDDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286451.1 | NP_001273380.1 | 3 | 408 | ||
NM_198527.3 | NP_940929.1 | 3 | 408 | |||
NR_104447.1 | 3 | 451 | ||||
GenBank mRNA | AK057584.1 | - | 1 | 829 | ||
BC033794.1 | - | 3 | 408 | 5122164 | ||
BQ777217.1 | - | 3 | 358 | |||
CV573934.1 | - | 3 | 393 |
