siRNA Details
| Gene Symbol: B3GNT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198540.2 | NP_940942.1 | 3 | 873 | ||
| XM_011526934.2 | XP_011525236.1 | 2 | 2192 | |||
| GenBank mRNA | AB175895.1 | - | 1 | 419 | ||
| AJ419172.1 | - | 1 | 419 | |||
| AY277592.2 | - | 3 | 873 | |||

B3GNT8
Genomic Map