siRNA Details
| Gene Symbol: NEXN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103535.1 | 1, 2 | 82 | |||
| GenBank mRNA | AK075118.1 | - | 1, 2 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEXN-AS1| Gene Symbol: NEXN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103535.1 | 1, 2 | 82 | |||
| GenBank mRNA | AK075118.1 | - | 1, 2 | 84 | ||