siRNA Details
| Gene Symbol: PFN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199346.2 | NP_955378.1 | 3, 4 | 638 | ||
| XM_005264292.2 | XP_005264349.1 | 4, 5 | 707 | |||
| XM_006712007.3 | XP_006712070.1 | 4, 5 | 449 | |||
| XM_011532823.2 | XP_011531125.1 | 4, 5 | 666 | |||
| GenBank mRNA | AK292601.1 | - | 3, 4 | 638 | ||
| BC029523.1 | - | 3, 4 | 442 | 5297707 | ||

PFN4
Genomic Map