siRNA Details
| Gene Symbol: FAM228B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145710.1 | NP_001139182.1 | 6 | 542 | ||
| NM_001291328.1 | NP_001278257.1 | 6 | 430 | |||
| GenBank mRNA | BC029459.1 | - | 6 | 422 | ||
| CN395744.1 | - | 6 | 554 | |||

FAM228B
Genomic Map