siRNA Details
| Gene Symbol: TMEM110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198563.2 | NP_940965.1 | 5 | 653 | ||
| GenBank mRNA | AY726591.1 | - | 5 | 417 | ||
| BC022209.1 | - | 3 | 280 | |||
| BC047015.1 | - | 5 | 603 | 5199769 | ||
| BX360795.2 | - | 5 | 668 | |||
| Gene Symbol: TMEM110-MUSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198974.2 | NP_001185903.2 | 5 | 653 | ||

TMEM110
Genomic Map