siRNA Details
| Gene Symbol: C5orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198566.3 | NP_940968.1 | 9 | 1792 | ||
| XM_011514038.2 | XP_011512340.1 | 9 | 1701 | |||
| XM_017009445.1 | XP_016864934.1 | 8 | 1823 | |||
| XM_017009446.1 | XP_016864935.1 | 7 | 1733 | |||
| GenBank mRNA | AK056925.1 | - | 9 | 1707 | ||
| BC036867.1 | - | 9 | 1655 | 5198039 | ||

C5orf34
Genomic Map