siRNA Details
| Gene Symbol: VWC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198570.4 | NP_940972.2 | 2 | 1231 | ||
| NR_136188.1 | 1 | 778 | ||||
| GenBank mRNA | AB292671.1 | - | 1 | 675 | ||
| AY358393.1 | - | 2 | 1255 | |||
| BC110857.1 | - | 2 | 1174 | 5731863 | ||

VWC2
Genomic Map