siRNA Details
| Gene Symbol: VWC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198570.4 | NP_940972.2 | 4 | 1516 | ||
| NR_136188.1 | 3 | 1063 | ||||
| GenBank mRNA | AB292671.1 | - | 3 | 960 | ||
| AY358393.1 | - | 4 | 1540 | |||
| BC110857.1 | - | 4 | 1459 | 5731863 | ||

VWC2
Genomic Map