siRNA Details
| Gene Symbol: SPATC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134374.1 | NP_001127846.1 | 4 | 1544 | ||
| NM_198572.2 | NP_940974.2 | 5 | 1684 | |||
| XM_011517021.1 | XP_011515323.1 | 6 | 1789 | |||
| XM_011517022.2 | XP_011515324.1 | 6 | 2075 | |||
| XM_011517023.2 | XP_011515325.1 | 6 | 3093 | |||
| XM_011517024.2 | XP_011515326.1 | 7 | 2314 | |||
| XM_011517025.2 | XP_011515327.1 | 6 | 2749 | |||
| XM_017013399.1 | XP_016868888.1 | 7 | 1608 | |||
| GenBank mRNA | AB092352.1 | - | 5 | 1672 | ||
| AK301710.1 | - | 4 | 1532 | |||
| BC050390.1 | - | 5 | 1674 | 5742710 | ||
| BC053547.1 | - | 5 | 1659 | 5743874 | ||

SPATC1
Genomic Map