siRNA Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198686.2 | NP_941959.1 | 4, 5 | 534 | ||
| GenBank mRNA | AB678452.1 | - | 4, 5 | 534 | ||
| BC040679.2 | - | 4, 5 | 457 | 4817835 | ||
| BX248046.1 | - | 3, 4 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB15| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198686.2 | NP_941959.1 | 4, 5 | 534 | ||
| GenBank mRNA | AB678452.1 | - | 4, 5 | 534 | ||
| BC040679.2 | - | 4, 5 | 457 | 4817835 | ||
| BX248046.1 | - | 3, 4 | 330 | |||