siRNA Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308154.1 | NP_001295083.1 | 1 | 93 | ||
| NM_198686.2 | NP_941959.1 | 1 | 93 | |||
| XM_017021277.1 | XP_016876766.1 | 1 | 133 | |||
| GenBank mRNA | AB672631.2 | - | 1 | 62 | ||
| AB672632.1 | - | 1 | 63 | |||
| AB678452.1 | - | 1 | 93 | |||
| AB678453.1 | - | 1 | 76 | |||
| BC040679.2 | - | 1 | 16 | 4817835 | ||
| CN352234.1 | - | 1 | 94 | |||

RAB15
Genomic Map