siRNA Details
| Gene Symbol: KLHL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198582.3 | NP_940984.3 | 2 | 290 | ||
| GenBank mRNA | AK125364.1 | - | 1 | 162 | ||
| BC103703.1 | - | 1 | 160 | |||
| BC104749.1 | - | 1 | 160 | |||
| BC105696.1 | - | 1 | 160 | |||
| BX280405.1 | - | 2 | 290 | |||

KLHL30
Genomic Map