siRNA Details
| Gene Symbol: KLHL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198582.3 | NP_940984.3 | 3, 4 | 1025 | ||
| GenBank mRNA | AK125364.1 | - | 2, 3 | 897 | ||
| BC103703.1 | - | 2, 3 | 895 | |||
| BC104749.1 | - | 2, 3 | 895 | |||
| BC105696.1 | - | 2, 3 | 895 | |||

KLHL30
Genomic Map