siRNA Details
| Gene Symbol: GPR153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207370.2 | NP_997253.2 | 5 | 1402 | ||
| XM_011541434.2 | XP_011539736.1 | 5 | 1506 | |||
| XM_017001250.1 | XP_016856739.1 | 4 | 1488 | |||
| XM_017001251.1 | XP_016856740.1 | 5 | 1391 | |||
| GenBank mRNA | BC068275.1 | - | 5 | 1402 | 5167285 | |

GPR153
Genomic Map