siRNA Details
| Gene Symbol: LRRC37A6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003525.2 | 1 | 2364 | |||
| GenBank mRNA | AK126872.1 | - | 1 | 620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC37A6P| Gene Symbol: LRRC37A6P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003525.2 | 1 | 2364 | |||
| GenBank mRNA | AK126872.1 | - | 1 | 620 | ||