siRNA Details
| Gene Symbol: CLEC2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130711.1 | NP_001124183.1 | 3 | 241 | ||
| NM_207375.2 | NP_997258.1 | 3 | 241 | |||
| XM_011520654.2 | XP_011518956.1 | 3 | 290 | |||
| XM_011520656.2 | XP_011518958.1 | 3 | 290 | |||
| XM_011520657.2 | XP_011518959.1 | 3 | 289 | |||
| GenBank mRNA | AY359126.1 | - | 3 | 207 | ||
| EF127467.1 | - | 3 | 241 | |||

CLEC2A
Genomic Map