siRNA Details
| Gene Symbol: SHISA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007538.1 | NP_001007539.1 | 2 | 934 | ||
| GenBank mRNA | AY358775.1 | - | 2 | 922 | ||
| BC132898.1 | - | 2 | 901 | 40146920 | ||
| BC137092.1 | - | 2 | 901 | 9021089 | ||

SHISA2
Genomic Map