siRNA Details
| Gene Symbol: TMEM179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286389.1 | NP_001273318.1 | 2 | 385 | ||
| NM_001286390.1 | NP_001273319.1 | 2 | 385 | |||
| XM_011536745.2 | XP_011535047.1 | 3 | 1112 | |||
| XM_011536750.2 | XP_011535052.1 | 3 | 297 | |||
| GenBank mRNA | AI299168.1 | - | 2 | 260 | ||
| AK124477.1 | - | 2 | 385 | |||
| AW026835.1 | - | 2 | 260 | |||

TMEM179
Genomic Map