siRNA Details
| Gene Symbol: LRRC75A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113567.2 | NP_001107039.1 | 2 | 530 | ||
| NM_207387.3 | NP_997270.2 | 2 | 530 | |||
| XM_011523845.2 | XP_011522147.1 | 2 | 451 | |||
| XM_017024619.1 | XP_016880108.1 | 2 | 299 | |||
| XM_017024620.1 | XP_016880109.1 | 3 | 462 | |||
| GenBank mRNA | AK093015.1 | - | 2 | 189 | ||
| BC144381.1 | - | 2 | 249 | |||
| DN997688.1 | - | 2 | 350 | |||

LRRC75A
Genomic Map