siRNA Details
Gene Symbol: AATK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027255.1 | 3 | 553 | |||
GenBank mRNA | AK126811.1 | - | 3 | 553 | ||
BC140737.1 | - | 3 | 544 | |||
BC171777.1 | - | 3 | 544 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AATK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027255.1 | 3 | 553 | |||
GenBank mRNA | AK126811.1 | - | 3 | 553 | ||
BC140737.1 | - | 3 | 544 | |||
BC171777.1 | - | 3 | 544 |