siRNA Details
| Gene Symbol: ZNF470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001668.3 | NP_001001668.3 | 6 | 1112 | ||
| XM_017026801.1 | XP_016882290.1 | 7 | 1186 | |||
| GenBank mRNA | AY484591.1 | - | 6 | 628 | ||
| BC136762.1 | - | 6 | 624 | 9020752 | ||
| BX641145.1 | - | 1 | 75 | |||

ZNF470
Genomic Map