siRNA Details
| Gene Symbol: RNF207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207396.2 | NP_997279.2 | 2 | 291 | ||
| XM_011541439.2 | XP_011539741.1 | 2 | 945 | |||
| XM_017001259.1 | XP_016856748.1 | 2 | 245 | |||
| GenBank mRNA | AK128246.1 | - | 3 | 851 | ||
| BC119780.1 | - | 2 | 339 | |||
| CD673733.1 | - | 2 | 291 | |||
| DB290694.1 | - | 2 | 247 | |||

RNF207
Genomic Map