siRNA Details
| Gene Symbol: CD164L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207397.3 | NP_997280.2 | 3 | 402 | ||
| XM_005245868.3 | XP_005245925.2 | 3 | 384 | |||
| XM_011541441.1 | XP_011539743.1 | 3 | 384 | |||
| GenBank mRNA | AY358761.1 | - | 3 | 372 | ||
| BC137466.1 | - | 3 | 361 | 9021466 | ||
| DB114521.1 | - | 3 | 403 | |||
| DB231017.1 | - | 3 | 384 | |||

CD164L2
Genomic Map