siRNA Details
| Gene Symbol: FAM69A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006605.4 | NP_001006606.2 | 1 | 126 | ||
| NM_001252269.1 | NP_001239198.1 | 1 | 126 | |||
| NM_001252270.1 | NP_001239199.1 | 1 | 126 | |||
| NM_001252273.1 | NP_001239202.1 | 1 | 126 | |||
| GenBank mRNA | AL536469.3 | - | 1 | 70 | ||
| AL702320.1 | - | 1 | 90 | |||
| BC070342.1 | - | 1 | 60 | 6498824 | ||
| BF979329.1 | - | 1 | 71 | |||
| DB199113.1 | - | 1 | 52 | |||
| DB475309.1 | - | 1 | 126 | |||
| DB498496.1 | - | 1 | 269 | |||

FAM69A
Genomic Map