siRNA Details
| Gene Symbol: LOC388882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110539.1 | 7 | 699 | |||
| GenBank mRNA | BC036910.1 | - | 7 | 699 | 5164270 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC388882| Gene Symbol: LOC388882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110539.1 | 7 | 699 | |||
| GenBank mRNA | BC036910.1 | - | 7 | 699 | 5164270 | |