siRNA Details
| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207404.3 | NP_997287.2 | 3, 4 | 505 | ||
| GenBank mRNA | AK127779.1 | - | 3, 4 | 505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF662| Gene Symbol: ZNF662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207404.3 | NP_997287.2 | 3, 4 | 505 | ||
| GenBank mRNA | AK127779.1 | - | 3, 4 | 505 | ||