siRNA Details
| Gene Symbol: GFRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207410.2 | NP_997293.2 | 8 | 1181 | ||
| GenBank mRNA | AY358198.1 | - | 8 | 1182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFRAL| Gene Symbol: GFRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207410.2 | NP_997293.2 | 8 | 1181 | ||
| GenBank mRNA | AY358198.1 | - | 8 | 1182 | ||